Methacryloyl chloride stabilised, Sigma-Aldrich®
Supplier: Merck
Stabilization:
Stabilised with hydroquinone monomethyl ether (4-methoxyphenol, MEHQ)
M-Clarity™ quality level = MQ200
523216-100ML
523216-1L
SIAL523216-100MLEA
165
EUR
SIAL523216-100ML
SIAL523216-1L
Methacryloyl chloride stabilised, Sigma-Aldrich®
Methacryloyl chloride
Formula:
C₄H₅ClO |
MDL Number:
MFCD00000716 CAS Number: 920-46-7 EINECS: 213-058-9 UN: 3383 ADR: 6.1,I |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...