Sodium hydroxymethanesulphinate dihydrate ≥98% (RT), Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Sodium hydroxymethanesulphinate dihydrate(Chunks or pellets), Hydroxymethanesulphinic acid sodium salt dihydrate, Formaldehyde sodium sulphoxylate dihydrate, Sodium formaldehydesulphoxylate hydrate, Rongalite dihydrate, Sodium formaldehydesulphoxylate dihydrate
71530-1KG 71530-5KG 71530-250G
SIAL71530-1KGEA 87.8 EUR
SIAL71530-1KG SIAL71530-5KG SIAL71530-250G
Sodium hydroxymethanesulphinate dihydrate ≥98% (RT), Sigma-Aldrich®
Sodium hydroxymethanesulphinate dihydrate
Formula: CH₃NaO₃S·2H₂O
MW: 154.12 g/mol
MDL Number: MFCD00150598
CAS Number: 6035-47-8
EINECS: 205-739-4