Disodium hydrogenarsenate heptahydrate, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
Synonyms:
Sodium hydrogen arsenate heptahydrate, Sodium arsenate dibasic heptahydrate, Arsenic Acid, Sodium Salt Heptahydrate. Sodium Salt
M-Clarity™ quality level = MQ200
A6756-100G
A6756-50G
SIALA6756-100GEA
146
EUR
SIALA6756-100G
SIALA6756-50G
Disodium hydrogenarsenate heptahydrate, Sigma-Aldrich®
Disodium hydrogenarsenate heptahydrate
Formula:
HAsNa₂O₄·7H₂O MW: 312.01 g/mol Melting Pt: 57 Density: 1.88 g/cm³ (20 °C) |
MDL Number:
MFCD00149179 CAS Number: 10048-95-0 EINECS: 231-902-4 UN: 1685 ADR: 6.1 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...