Cystamine dihydrochloride, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
C121509-25G C121509-100G C121509-500G
SIALC121509-25GEA 39.1 EUR
SIALC121509-25G SIALC121509-100G SIALC121509-500G
Cystamine dihydrochloride, Sigma-Aldrich®
Cystamine dihydrochloride
Formula: C₄H₁₂N₂S₂·2HCl
MDL Number: MFCD00012905
CAS Number: 56-17-7
EINECS: 200-260-7