Orthoboric acid ≥99.99% (metals basis)

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Boron trihydroxide, Boron hydroxide, Boric acid, Boracic acid

36771.36 36771.A3
36771.36EA 71.6 EUR
36771.36 36771.A3
Orthoboric acid ≥99.99% (metals basis)
ortho-Boric acid

Purity: 99.99% CAS No.: 10043-35-3 Molecular Formula: BH3O3 Formula Weight: 61.83 Flash Point: None Physical Form: +8 Mesh Granular Density: 1.435 Appearance: White Odour: Odourless pH: 3.8-4.8 Melting Point: ca 185° dec. Beilstein Reference: 1697939 Merck Reference: 14,1336 MDL No.: MFCD00011337

Formula: B(OH)₃
MW: 61.83 g/mol
Boiling Pt: 185
Melting Pt: ∼ 185 °C (dec.)
Density: 1.435
Storage Temperature: Ambient
MDL Number: MFCD00011337
CAS Number: 10043-35-3
EINECS: 233-139-2
REACH: 01-2119486683-25
Merck Index: 14,1336
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=098dd700154daaf6338ebb270902bb2bc704f27d&vid=6eec7d529090dd4c4b966ed8cd8d057153c5a47b&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR