Tetrachlorosilane, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Silicon tetrachloride, Silicon (IV) chloride
M-Clarity™ quality level = MQ200
215120-1KG 215120-100G
SIAL215120-1KGEA 119 EUR
SIAL215120-1KG SIAL215120-100G
Tetrachlorosilane, Sigma-Aldrich®
Tetrachlorosilane
Formula: Cl₄Si
MW: 169.9 g/mol
Boiling Pt: 57.6
Melting Pt: –70 °C
Density: 1.48 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011229
CAS Number: 10026-04-7
EINECS: 233-054-0
UN: 1818
ADR: 8 II
Merck Index: 12,08644