Silica gel, Pore size: 60 Å 0.019-0.037 mm (400-600 mesh)
Supplier: Thermo Fisher Scientific
Warning
42729.18 42729.30 42729.A1 42729.A7
42729.18EA 45.6 EUR
42729.18 42729.30 42729.A1 42729.A7
Silica gel, Pore size: 60 Å 0.019-0.037 mm (400-600 mesh)
Silica gel
CAS No.: 7631-86-9 Physical Form: -400+600 Mesh Powder, S.A. 500-600m2/g Odour: Odourless MDL No.: MFCD00011232
Column chromatography with elevated pressure
Formula: SiO₂
Storage Temperature: Ambient
MDL Number: MFCD00011232
CAS Number: 63231-67-4
EINECS: 231-545-4