Phosphorus(III) bromide ≥98%
Supplier: Thermo Fisher Scientific
Danger
L13638.14 L13638.22 L13638.0B
L13638.14EA 38.5 EUR
L13638.14 L13638.22 L13638.0B
Phosphorus(III) bromide ≥98%
Phosphorus (III) bromide
Formula: PBr₃
Boiling Pt: 175 °C (1013 hPa)
Melting Pt: –40 °C
Density: 2.8903 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011436
CAS Number: 7789-60-8
EINECS: 232-178-2
UN: 1808
ADR: 8,II
Merck Index: 12,07514