7-Methyl-4-indanol

Supplier: Thermo Scientific
Warning

Synonyms: 7-Methyl-2,3-dihydro-1H-inden-4-ol, 7-Methylindan-4-ol

BTB09366DA BTB09366EA BTB09366.50
MAYBBTB09366DAEA 0 EUR
MAYBBTB09366DA MAYBBTB09366EA MAYBBTB09366.50
7-Methyl-4-indanol
7-Methyl-4-indanol
Formula: C₁₀H₁₂O
MDL Number: MFCD00173675
CAS Number: 16400-13-8
EINECS: 240-450-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR