Silver nitrate
Supplier: BIOMOL RESEARCH LABORATORIES
Danger
Synonyms: Lunar caustic, Silver mononitrate, Nitric acid silver salt
50409-1 50409-2
BMOL50409-1EA 0 EUR
BMOL50409-1 BMOL50409-2
Silver nitrate
Silver nitrate
Formula: AgNO₃
MW: 169.87 g/mol
Boiling Pt: 444
Melting Pt: 210
Density: 4.35 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003414
CAS Number: 7761-88-8
EINECS: 231-853-9
UN: 1493
ADR: 5.1 II
REACH: 01-2119513705-43
Merck Index: 12,08661
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f56d5ff35f4c0937041d9629e42aae69bf1987b3&vid=7ca52a55b378bddc27fb4a51ace3ae27a0451c75&prid=web&clid=SAPDAM