Osmium tetraoxide
Supplier: Roth Carl
Danger
8371.1 8371.2 8371.3
ROTH8371.1EA 68.5 EUR
ROTH8371.1 ROTH8371.2 ROTH8371.3
Osmium tetraoxide
Osmium tetraoxide
Formula: O₄Os
Boiling Pt: 130 °C (1013 hPa)
Melting Pt: 40 °C
Density: 1.04 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011150
CAS Number: 20816-12-0
EINECS: 244-058-7
UN: 2471
ADR: 6.1,I
Merck Index: 13,06962