1-Chloronaphthalene, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Triisodecyl phosphite, α-Chloronaphthalene
M-Clarity™ quality level = MQ100
185752-500G 185752-100G
SIAL185752-500GEA 114 EUR
SIAL185752-500G SIAL185752-100G
1-Chloronaphthalene, Sigma-Aldrich®
1-Chloronaphthalene
Formula: C₁₀H₇Cl
Boiling Pt: 263 °C (1013 hPa)
Melting Pt: –2.5 °C
Density: 1.1938 g/cm³ (20 °C)
Flash Pt: 121 °C
Storage Temperature: Ambient
MDL Number: MFCD00003874
CAS Number: 90-13-1
EINECS: 201-967-3
UN: 2810
ADR: 6.1,III
Merck Index: 13,02168