1,4-Dimethoxynaphthalene 96%
Supplier: Thermo Fisher Scientific
|
Warning
|
A14095.03
A14095.06
A14095.14
AB116050.0005
A14095.03EA
79.6
EUR
A14095.03
A14095.06
A14095.14
ABCRAB116050.0005
1,4-Dimethoxynaphthalene 96%
1,4-Dimethoxynaphthalene
Purity: 96%
CAS No.: 10075-62-4
Molecular Formula: C12H12O2
Formula Weight: 188.23
Melting Point: 84-85°
Beilstein Reference: 2048082
Formula:
C₁₂H₁₂O₂ Melting Pt: 84…85 °C Storage Temperature: Ambient |
MDL Number:
MFCD00052378 CAS Number: 10075-62-4 EINECS: 233-209-2 |