2,3-Dimethylnaphthalene ≥97%
Supplier: Thermo Fisher Scientific
Warning
L08386.03 L08386.06
L08386.03EA 69.9 EUR
L08386.03 L08386.06
2,3-Dimethylnaphthalene ≥97%
2,3-Dimethylnaphthalene
Purity: 97% CAS No.: 581-40-8 Molecular Formula: C12H12 Formula Weight: 156.23 Melting Point: 103-106° Boiling Point: 265-268° MDL No.: MFCD00004119
Formula: C₁₂H₁₂
MW: 156.23 g/mol
Boiling Pt: 265…268 °C
Melting Pt: 103…106 °C
Density: 1.003 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004119
CAS Number: 581-40-8
EINECS: 209-463-5