1-Phenylnaphthalene ≥97%
Supplier: Thermo Fisher Scientific
B21953.06
B21953.14
B21953.22
B21953.06EA
59.3
EUR
B21953.06
B21953.14
B21953.22
1-Phenylnaphthalene ≥97%
1-Phenylnaphthalene
Purity: 97%
CAS No.: 605-02-7
Molecular Formula: C16H12
Formula Weight: 204.27
Flash Point:
Formula:
C₁₆H₁₂ Boiling Pt: 320 °C Melting Pt: 45 °C Density: 1.088 Flash Pt: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003983 CAS Number: 605-02-7 EINECS: 210-081-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...