General purpose gloves, Perfect Poly® Finger
Supplier: HONEYWELL SAFETY
2232240/10 2232240/7 2232240/6 2232240/8 2232240/9
DALL2232240/10EA 25.2 EUR
DALL2232240/10 DALL2232240/7 DALL2232240/6 DALL2232240/8 DALL2232240/9
General purpose gloves, Perfect Poly® Finger
Gloves
White polyamide knitted glove with white polyurethane coating on fingertips and an elastic wrist.
  • Tight knit gives the glove perfect support, limits the penetration of dirt and offers a good touch
  • Ventilated back and palm optimise the glove's breathability
  • PU coating ensures a good grip at the fingertips
  • Non pilling: Polyamide is suitable for precision work
Suitable for handling fine products in a dry environment. It is certified as 'silicone-free' to ensure its use in painting environments.
Certifications: Cat. I