2,6-Dibromopyridine
Supplier: Thermo Scientific
SB01233DA SB01233.10 SB01233.50
MAYBSB01233DAEA 0 EUR
MAYBSB01233DA MAYBSB01233.10 MAYBSB01233.50
2,6-Dibromopyridine
2,6-Dibromopyridine
Formula: C₅H₃Br₂N
Boiling Pt: 255 °C (1013 hPa)
Melting Pt: 116…119 °C
Storage Temperature: Ambient
MDL Number: MFCD00006223
CAS Number: 626-05-1
EINECS: 210-926-9
UN: 2811
ADR: 6.1,II