2,3,5,6-Tetrachloropyridine ≥98%
Supplier: Thermo Fisher Scientific
|
Danger
|
L19458.03
L19458.06
L19458.14
L19458.03EA
14.6
EUR
L19458.03
L19458.06
L19458.14
2,3,5,6-Tetrachloropyridine ≥98%
2,3,5,6-Tetrachloropyridine
Purity: 98%
CAS No.: 2402-79-1
Molecular Formula: C5HCl4N
Formula Weight: 216.88
Flash Point: 188°(370°F)
Melting Point: 89-93°
Boiling Point: 250-252°
Beilstein Reference: 129639
UN No.: 3077
MDL No.: MFCD00092428
Formula:
C₅HCl₄N Boiling Pt: 250…252 °C Melting Pt: 89…93 °C Flash Pt: 188 °C (370 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00092428 CAS Number: 2402-79-1 EINECS: 219-283-9 UN: 3077 ADR: 9,III |