3-Bromothiophene
Supplier: Thermo Scientific
Synonyms: 3-Thienyl bromide
CC13110.1 CC13110.10 CC13110.50
MAYBCC13110.1EA 0 EUR
MAYBCC13110.1 MAYBCC13110.10 MAYBCC13110.50
3-Bromothiophene
3-Bromothiophene
Formula: C₄H₃BrS
MW: 163.04 g/mol
Boiling Pt: 158
Melting Pt: –10 °C
Density: 1.735 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00005464
CAS Number: 872-31-1
EINECS: 212-821-3
UN: 2810
ADR: 6.1 II