2-Methylthiophene 98%
Supplier: Thermo Fisher Scientific
Danger
127850250 127851000
ACRO127850250EA 35.4 EUR
ACRO127850250 ACRO127851000
2-Methylthiophene 98%
2-Methylthiophene
Formula: C₅H₆S
MW: 98.17 g/mol
Boiling Pt: 113 °C (760 mmHg)
Melting Pt: –63 °C
Density: 1.02 g/cm³
Flash Pt: 7 °C
Storage Temperature: Ambient
MDL Number: MFCD00005451
CAS Number: 554-14-3
EINECS: 209-063-0
UN: 1993
ADR: 3 II
Order Now
Specification Test Results
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5190 to 1.5210 (20°C, 589 nm)
GC ≥97.5 %