Tris(2-ethylhexyl)phosphate, Sigma-Aldrich®
Supplier: Merck
Synonyms: Trioctyl phosphate, Phosphoric acid tris(2-ethylhexyl ester), Tris(2-ethylhexyl) phosphate
M-Clarity™ quality level = MQ200
289922-25ML 289922-500ML
SIAL289922-25MLEA 73.4 EUR
SIAL289922-25ML SIAL289922-500ML
Tris(2-ethylhexyl)phosphate, Sigma-Aldrich®
Tris(2-ethylhexyl) phosphate
Formula: C₂₄H₅₁O₄P
MDL Number: MFCD00009491
CAS Number: 78-42-2
EINECS: 201-116-6