Lithium azide 20% in water, pure
Supplier: Thermo Fisher Scientific
Danger
335360250 335361000
ACRO335360250EA 136 EUR
ACRO335360250 ACRO335361000
Lithium azide 20% in water, pure
Lithium azide
Formula: LiN₃
MW: 48.96 g/mol
Boiling Pt: 103
Density: 1.83
Storage Temperature: Ambient
MDL Number: MFCD00041956
CAS Number: 19597-69-4
EINECS: 243-177-1
UN: 3287
ADR: 6.1 I
Order Now
Specification Test Results
Appearance Clear colorless Solution
Titration Argentometric 17 to 23 %