1-Aminobenzotriazole, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
A3940-250MG A3940-50MG
SIALA3940-250MGEA 226 EUR
SIALA3940-250MG SIALA3940-50MG
1-Aminobenzotriazole, Sigma-Aldrich®
1-Aminobenzotriazole
Formula: C₆H₆N₄
MDL Number: MFCD00132902
CAS Number: 1614-12-6