5-Aminobenzotriazole 96%
Supplier: Thermo Fisher Scientific
|
Warning
|
L06660.03
L06660.06
L06660.03EA
102
EUR
L06660.03
L06660.06
5-Aminobenzotriazole 96%
5-Aminobenzotriazole
Purity: 98%
CAS No.: 3325-11-9
Molecular Formula: C6H6N4
Formula Weight: 134.14
Odour: Odourless
Melting Point: 157-162°
Beilstein Reference: 121262
MDL No.: MFCD00047209
Formula:
C₆H₆N₄ Melting Pt: 157…162 °C Storage Temperature: Ambient |
MDL Number:
MFCD00047209 CAS Number: 3325-11-9 |