Oxalyl dichloride 2.0 M in dichloromethane, AcroSeal®
Supplier: Thermo Fisher Scientific
Danger
Synonyms: Oxalyl chloride
369181000 369188000
369181000.EA 51.7 EUR
369181000. 369188000.
Oxalyl dichloride 2.0 M in dichloromethane, AcroSeal®
Oxalyl dichloride
Formula: C₂Cl₂O₂
MW: 126.93 g/mol
Boiling Pt: 63
Melting Pt: -12
Storage Temperature: Refrigerator
MDL Number: MFCD00000704
CAS Number: 79-37-8
EINECS: 201-200-2
UN: 2922
ADR: 8 II
Merck Index: 12,07046
Order Now
Specification Test Results
Appearance Clear colorless to yellow Solution
Titration Argentometric 1.9 to 2.1 M