Reactions vessels, micro, BOLA
Supplier: Bohlender
B318-80 B318-64 B318-40
BOHLB318-80EA 944 EUR
BOHLB318-80 BOHLB318-64 BOHLB318-40
Reactions vessels, micro, BOLA
Reaction Vessels Reaction Vessels, Beaker Shape
PFA, translucent.
  • Temperature resistance from −200 to +250°C
  • Very good chemical resistance
  • FDA conform
Vessel made of translucent, non-porous PFA, screw cap made of PTFE with threaded necks for connection of e.g. thermometers, probes or sensors.