Ethidium bromide 10 mg/ml in aqueous solution, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: 3,8-Diamino-5-ethyl-6-phenylphenanthridinium bromide, 3,8-Diamino-5-ethyl-6-phenylphenanthridin-5-ium bromide
46067-250ML-F 46067-50ML-F
SIAL46067-250ML-FEA 408 EUR
SIAL46067-250ML-F SIAL46067-50ML-F
Ethidium bromide 10 mg/ml in aqueous solution, Sigma-Aldrich®
Ethidium bromide
Formula: C₂₁H₂₀BrN₃
MW: 394.31 g/mol
Boiling Pt: 72
MDL Number: MFCD00011724
CAS Number: 1239-45-8
EINECS: 214-984-6
Merck Index: 13,04751