Magnesium chloride 1 mol/l (2 N) in aqueous solution, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
63069-100ML 63069-500ML
SIAL63069-100MLEA 127 EUR
SIAL63069-100ML SIAL63069-500ML
Magnesium chloride 1 mol/l (2 N) in aqueous solution, Sigma-Aldrich®
Magnesium chloride
Formula: MgCl₂
Storage Temperature: Ambient
MDL Number: MFCD00011106
CAS Number: 7786-30-3
EINECS: 232-094-6
Merck Index: 13,05684