Pyridoxal hydrochloride, Sigma-Aldrich®
Supplier: Merck
Synonyms: 3-Hydroxy-5-(hydroxymethyl)-2-methylisonicotinaldehyde hydrochloride, Pyridoxal HCl
M-Clarity™ quality level = MQ200
P6155-25G P6155-5G
SIALP6155-25GEA 632 EUR
SIALP6155-25G SIALP6155-5G
Pyridoxal hydrochloride, Sigma-Aldrich®
Pyridoxal hydrochloride
Formula: C₈H₁₀ClNO₃
MW: 203.62 g/mol
MDL Number: MFCD00012809
CAS Number: 65-22-5
EINECS: 200-602-5