Chlorohydroquinone 90% tech.
Supplier: Thermo Fisher Scientific
|
Danger
|
208220050
208221000
ACRO208220050EA
20
EUR
ACRO208220050
ACRO208221000
Chlorohydroquinone 90% tech.
Chlorohydroquinone
Formula:
C₆H₅ClO₂ Melting Pt: 101…102 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002341 CAS Number: 615-67-8 EINECS: 210-442-8 ADR: 6.1,III |
Specification Test Results
Appearance | Grey to light brown Crystalline powder |
Infrared spectrum | Conforms |
GC | ≥88 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...