Dextran, M.W. ~ 20,000 (from Leuconostoc sp.), BioChemika®, Sigma-Aldrich®
Supplier: Merck
31387-100G 31387-25G 31387-500G
SIAL31387-100GEA 158 EUR
SIAL31387-100G SIAL31387-25G SIAL31387-500G
Dextran, M.W. ~ 20,000 (from Leuconostoc sp.), BioChemika®, Sigma-Aldrich®
Dextran
Formula: (C₆H₁₀O₅)n
MDL Number: MFCD00130935
CAS Number: 9004-54-0
EINECS: 232-677-5