Dilithium tetraborate trihydrate 96%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: Lithium tetraborate trihydrate
A10358.22 A10358.36
A10358.22EA 36.6 EUR
A10358.22 A10358.36
Dilithium tetraborate trihydrate 96%
di-Lithium tetraborate trihydrate
Purity: 96% CAS No.: 37195-62-3 Molecular Formula: Li2B4O7.3H2O Formula Weight: 223.17 (169.12anhy) Merck Reference: 14,5524 MDL No.: MFCD00167253
Formula: B₄Li₂O₇·3H₂O
MW: 223.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00167253
CAS Number: 37195-62-3
EINECS: 234-514-3
Merck Index: 14,5524