Zinc(II) nitrate hydrate, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ100
230006-250G 230006-25G
SIAL230006-250GEA 755 EUR
SIAL230006-250G SIAL230006-25G
Zinc(II) nitrate hydrate, Sigma-Aldrich®
Zinc (II) nitrate hydrate
Formula: N₂O₆Zn·H₂O
MW: 207.42 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00149888
CAS Number: 13778-30-8
EINECS: 231-943-8
UN: 1514
ADR: 5.1 II