4-Amino-2,2,6,6-tetramethylpiperidinooxy (4-amino-TEMPO) 97% free radical
Supplier: Thermo Fisher Scientific
Danger
Synonyms: 4-Amino-2,2,6,6-tetramethylpiperidine 1-Oxyl, 4-Amino-1-oxy-2,2,6,6-tetramethylpiperidine
172730010 172730050 172732500
ACRO172730010EA 143 EUR
ACRO172730010 ACRO172730050 ACRO172732500
4-Amino-2,2,6,6-tetramethylpiperidinooxy (4-amino-TEMPO) 97% free radical
4-Amino-2,2,6,6-tetramethylpiperidinooxy (4-Amino-TEMPO, free radical)
Formula: C₉H₁₉N₂O
MW: 171.26 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00006479
CAS Number: 14691-88-4
EINECS: 238-738-2
UN: 3263
ADR: 8 III
Order Now
Specification Test Results
Appearance Red to red-brown Adhering crystals or needles or moist mass
Infrared spectrum Conforms
GC ≥96.0 %