Ammonium tetrachlorocuprate dihydrate, Sigma-Aldrich®
Supplier: Merck
Synonyms: Ammonium copper(II) chloride dihydrate
M-Clarity™ quality level = MQ100
00634-1KG 00634-250G
SIAL00634-1KGEA 135 EUR
SIAL00634-1KG SIAL00634-250G
Ammonium tetrachlorocuprate dihydrate, Sigma-Aldrich®
Ammonium tetrachlorocuprate dihydrate
Formula: H₈Cl₄CuN₂·2H₂O
MW: 277.46 g/mol
MDL Number: MFCD00150384
CAS Number: 10060-13-6
EINECS: 239-690-5