Pyruvaldehyde (methylglyoxal) 35 - 45% (w/w) in aqueous solution
Supplier: Thermo Fisher Scientific
Danger
Synonyms: Pyruvicaldehyde, Pyruvic aldehyde
B24664.14 B24664.22 B24664.36
B24664.14EA 34.5 EUR
B24664.14 B24664.22 B24664.36
Pyruvaldehyde (methylglyoxal) 35 - 45% (w/w) in aqueous solution
Pyruvaldehyde (Methylglyoxal)
CAS No.: 78-98-8 Molecular Formula: C3H4O2 Formula Weight: 72.06 Refractive Index: 1.4050 Beilstein Reference: 906750 Merck Reference: 14,6081 MDL No.: MFCD00006960
Formula: C₃H₄O₂
Density: 1.2
Storage Temperature: Ambient
MDL Number: MFCD00006960
CAS Number: 78-98-8
EINECS: 201-164-8
Merck Index: 14,6081