Potassium-tert-butoxide 1 mol/l in THF, Sigma-Aldrich®
Supplier: Merck
Synonyms: Potassium-2-methylpropan-2-olate, Potassium tert-butylate, Potassium tert-butanolate, Potassium t-butoxide
M-Clarity™ quality level = MQ200
328650-2L 328650-500ML 328650-50ML
SIAL328650-2LEA 846 EUR
SIAL328650-2L SIAL328650-500ML SIAL328650-50ML
Potassium-tert-butoxide 1 mol/l in THF, Sigma-Aldrich®
Potassium-tert-butoxide
Formula: C₄H₉KO
MW: 112.21 g/mol
MDL Number: MFCD00012162
CAS Number: 865-47-4
EINECS: 212-740-3