Tributyl acetylcitrate, Sigma-Aldrich®

Supplier: Merck

Synonyms: Tributyl O-Acetylcitrate, Acetyltributylcitrate, Acetyl tributyl citrate, Tributyl 2-acetoxypropane-1,2,3-tricarboxylate, O-Acetylcitric acid tributyl ester

M-Clarity™ quality level = MQ300

W308005-1KG W308005-5KG W308005-SAMPLE
SIALW308005-1KGEA 75.1 EUR
SIALW308005-1KG SIALW308005-5KG SIALW308005-SAMPLE
Tributyl acetylcitrate, Sigma-Aldrich®
Tributyl acetylcitrate
Formula: C₂₀H₃₄O₈
MDL Number: MFCD00043554
CAS Number: 77-90-7
EINECS: 201-067-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR