(3-Acryloxypropyl)trimethoxysilane, Sigma-Aldrich®

Supplier: Merck

Synonyms: 3-(Trimethoxysilyl)propyl acrylate

M-Clarity™ quality level = MQ100

475149-25ML 475149-5ML
SIAL475149-25MLEA 469 EUR
SIAL475149-25ML SIAL475149-5ML
(3-Acryloxypropyl)trimethoxysilane, Sigma-Aldrich®
(3-Acryloxypropyl)trimethoxysilane
Formula: C₉H₁₈O₅Si
MW: 234.32 g/mol
MDL Number: MFCD00054803
CAS Number: 4369-14-6
EINECS: 419-560-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR