Disodium fumarate, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ200

F1506-100G F1506-25G F1506-500G
SIALF1506-100GEA 71.9 EUR
SIALF1506-100G SIALF1506-25G SIALF1506-500G
Disodium fumarate, Sigma-Aldrich®
Formula: C₄H₂Na₂O₄
MDL Number: MFCD00064567
CAS Number: 17013-01-3
EINECS: 241-087-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR