Bismuth(III) hydroxide, tech.
Supplier: Thermo Fisher Scientific
Warning
Synonyms: Bismuth hydroxide
40397.30 40397.A1
40397.30EA 129 EUR
40397.30 40397.A1
Bismuth(III) hydroxide, tech.
Bismuth (III) hydroxide
CAS No.: 10361-43-0 Molecular Formula: Bi(OH)3 Formula Weight: 260.00 Physical Form: Amorphous Powder Odour: Odourless Solubility: Insoluble in water. Soluble in acids MDL No.: MFCD00046188
Hydrolysis of ribonucleic acid, absorbent
Formula: H₃BiO₃
MW: 260 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00046188
CAS Number: 10361-43-0
EINECS: 233-790-2