2-(2-Bromoethyl)-1,3-dioxolane 95% stabilised
Supplier: Thermo Fisher Scientific
|
Danger
|
Stabilization:
Stabilised with silver
A11441.09
A11441.18
A11441.09EA
43.3
EUR
A11441.09
A11441.18
2-(2-Bromoethyl)-1,3-dioxolane 95% stabilised
2-(2-Bromoethyl)-1,3-dioxolane
Stabilized with silver
Purity: 95%
CAS No.: 18742-02-4
Molecular Formula: C5H9BrO2
Formula Weight: 181.04
Storage Temperature: 2°C to 8°C
Flash Point: 65°(149°F)
Refractive Index: 1.4790
Boiling Point: 68-70°/8mm
Beilstein Reference: 103516
UN No.: 2810
MDL No.: MFCD00003216
Formula:
C₅H₉BrO₂ Boiling Pt: 68…70 °C (8 mmHg) Density: 1.512 Flash Pt: 65 °C (149 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00003216 CAS Number: 18742-02-4 EINECS: 242-551-1 UN: 2810 ADR: 6.1,III |