4,5-Diphenyl-1,2,3-thiadiazole

Supplier: Thermo Scientific

SEW03000.1 SEW03000.10
MAYBSEW03000.1EA 0 EUR
MAYBSEW03000.1 MAYBSEW03000.10
4,5-Diphenyl-1,2,3-thiadiazole
4,5-Diphenyl-1,2,3-thiadiazole
Formula: C₁₄H₁₀N₂S
MDL Number: MFCD00082687
CAS Number: 5393-99-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR