Cytological fixative, CytoFixx™
Supplier: CELLPATH
NHB-0100-01A NHB-0101-01A NHB-0200-01A
720-2338EA 70.2 EUR
720-2338 720-2339 720-2340
Cytological fixative, CytoFixx™
Stains Microscopy and staining reagents
Ready-to-use solution of alcohol and carbowax, which gives similar fixation results and staining.