Bath fluids
Supplier: JULABO GmbH
8940111 8940114 8940115 8940125 8940100 8940101 8940103 8940104 8940105 8940107 8940109 8940124 8940102 8940130 8940133 8940132 8940138 8940139 8940140 8940141 8940108 8940106 8940142 8940143 8940113
461-3102EA 430 EUR
461-3102 461-3103 461-0226 461-3122 461-0111 461-0110 461-0120 461-0131 461-0130 461-0140 461-0225 461-0474 461-0121 461-0475 461-0478 461-0477 461-0875 461-0876 461-0877 461-0878 461-0151 461-0141 JULA8940142 JULA8940143 JULA8940113
Bath fluids
Circulator Bath Fluids
Bath fluids have been carefully selected and proven in long-term service. They are suitable for use with heating and refrigerated circulators whenever water cannot be employed as bath fluid. The choice of bath fluid is critical for best results in temperature control and operation of the circulator.
  • Low toxicity and almost odourless
  • High stability and good thermal conductivity
  • Low viscosity and low corrosiveness