Industrial Filter Papers
Supplier: Ahlstrom-Munksjö
200000 200001 200002 200003 200008 200012 200005 213156 200006
512-6131EA 113 EUR
512-6131 512-6130 512-6129 512-6128 512-6126 512-6124 512-6220 512-6198 512-6215
Industrial Filter Papers
Filters Filter Papers Qualitative Filter Paper
The following filter papers fulfill specific technical requirements corresponding to the requirements mentioned overleaf. Embossed filter papers have a larger surface area and, therefore, correspondingly shorter filtration times. They show a very homogenous and fine pore structure.
Grade 1200: Medium fast filtration
Grade 1300: Medium fast filtration for absorption, protection, oil filtration
Grade 1310: Medium fast filtration for absorption, protection, oil filtration
Grade 1350: Embossed, medium fast filtration for absorption, protection
Grade 1360: Medium fast filtration forMore Product Information
Order Now
SPECIFICATIONS
Grade 1300 1350 1360 17 1310
Environmentally Preferable
Weight 90 g/m² 90 g/m² 135 g/m² 95 g/m² 135 g/m²
Thickness 0,19 mm 0,25 mm 0,31 mm 0,28 mm 0,26 mm
Particle retention 7 - 10 µm 7 - 10 µm 7 - 10 µm 7 - 10 µm
Filtration speed 30 s (s/10 ml) 30 s (s/10 ml) 30 s (s/10 ml) 7 s (s/10 ml) 30 s (s/10 ml)