Conductivity and TDS standards
Supplier: Hanna
HI7033L HI70030P HI7034L/C
663-5054EA 24.8 EUR
663-5054 HANNHI70030P HANNHI7034L/C
Conductivity and TDS standards
Standards Conductivity Standards
An extensive range of standard solutions designed to ensure maximum accuracy of conductivity meters and sensors.
  • The labels on the bottles and sachets include the batch number, expiry date and the relationship between conductivity and temperature
The labels on the bottles and sachets include the batch number, expiry date and the relationship between conductivity and temperature.