VWR®, Pierceable Films for ELISA and General Incubation
Supplier: VWR Collection
391-1263EA 83.7 EUR
391-1263 391-1277
VWR®, Pierceable Films for ELISA and General Incubation
Seals Microplate Seals
Easily pierceable 94,1 µm thick PE films with acrylic adhesive allow direct sample recovery with singe- or multi-channel pipettes and robotic probes.
  • Functional temperature range from −40 to 90 °C
Each film L×D: 142,9×82,6 mm
Each film L×D: 142,9×82,6 mm