Real-Time PCR detection kits for microorganisms, QuickBlue RealQuick
Supplier: Q BIOANALYTIC
QB-RT-06-10SMP QB-RT-06-50 QB-RT-06-250 QB-RT-06-1000
710-0674EA 0 EUR
710-0674 710-0676 710-0675 710-0673
Real-Time PCR detection kits for microorganisms, QuickBlue RealQuick
Microbiology Test Systems PCR Tests
This product range is optimised for real-Time PCR and uses Taqman® probe technology for the detection of human pathogenic bacteria. The kits are typically co-ordinated in such a way as to enable them to be used together in one run.
  • Compatible with all major Real-Ttime PCR instruments
  • Excellent results with regard to specificity and sensitivity
  • In different packaging units (50, 250 and 1000 reactions) available
Certifications: Compliant with ISO standard 20838:2006.