Tris-Borate-EDTA (TBE) buffer, 10X
Supplier: PanReac AppliChem
A0972.1000
A0972.5000PE
A0972.1000EA
45.8
EUR
A0972.1000
A0972.5000PE
Tris-Borate-EDTA (TBE) buffer, 10X
Buffers
Electrophoresis Buffers
Running Buffers
10X concentrated solution of 0,89 M Tris, 0,89 M boric acid and 0,02 M EDTA.Na₂, pH 8,3 ±0,2.
CT = Cubitainer®